Anti-PPID Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302580EA 547 EUR
ORIGTA302580
Anti-PPID Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: PPID (peptidylprolyl isomerase D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR