Anti-RAD9A Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302588EA 550 EUR
ORIGTA302588
Anti-RAD9A Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR