Anti-RPS19 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302593EA 500 EUR
ORIGTA302593
Anti-RPS19 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: RPS19 (ribosomal protein S19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR