Anti-SAMSN1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302597EA 500 EUR
ORIGTA302597
Anti-SAMSN1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.5-1.5µg/ml

Type: Primary
Antigen: SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR