Anti-SLPI Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA306439EA 600 EUR
ORIGTA306439
Anti-SLPI Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: SLPI (secretory leukocyte peptidase inhibitor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR