Anti-MPP3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA308232EA 655 EUR
ORIGTA308232
Anti-MPP3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: MPP3 (membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR