Anti-CAMK1D Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA309027EA 681 EUR
ORIGTA309027
Anti-CAMK1D Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: CAMK1D (calcium/calmodulin-dependent protein kinase ID)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR