Anti-HSP90AB1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA309400EA 581 EUR
ORIGTA309400
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions:

Type: Primary
Antigen: HSP90AB1 (heat shock protein 90kDa alpha (cytosolic), class B member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR