Anti-ITGA11 Goat Antibody

Toimittaja: OriGene
ORIGTA311598EA 513 EUR
ORIGTA311598
Anti-ITGA11 Goat Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ITGA11 (integrin, alpha 11)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR