Anti-HTR4 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA314044EA 564 EUR
ORIGTA314044
Anti-HTR4 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: HTR4 (5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR