Anti-F2RL3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA314845EA 532 EUR
ORIGTA314845
Anti-F2RL3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: F2RL3 (coagulation factor II (thrombin) receptor-like 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR