Anti-GTF3C3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA314896EA 552 EUR
ORIGTA314896
Anti-GTF3C3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: GTF3C3 (general transcription factor IIIC, polypeptide 3, 102kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR