Anti-PSMD6 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA314977EA 552 EUR
ORIGTA314977
Anti-PSMD6 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: PSMD6 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR