Anti-SRSF8 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315126EA 552 EUR
ORIGTA315126
Anti-SRSF8 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: SRSF8 (serine/arginine-rich splicing factor 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR