Anti-PPRC1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315218EA 532 EUR
ORIGTA315218
Anti-PPRC1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PPRC1 (peroxisome proliferator-activated receptor gamma, coactivator-related 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR