Anti-LUC7L2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315490EA 552 EUR
ORIGTA315490
Anti-LUC7L2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: LUC7L2 (LUC7-like 2 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR