Anti-KCNK12 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315620EA 552 EUR
ORIGTA315620
Anti-KCNK12 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: KCNK12 (potassium channel, subfamily K, member 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR