Anti-ARID1B Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315663EA 552 EUR
ORIGTA315663
Anti-ARID1B Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:00-1:500; ELISA: 1:10000

Type: Primary
Antigen: ARID1B (AT rich interactive domain 1B (SWI1-like))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR