Anti-ZNF668 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315803EA 552 EUR
ORIGTA315803
Anti-ZNF668 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ZNF668 (zinc finger protein 668)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR