Anti-NIFK Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315914EA 552 EUR
ORIGTA315914
Anti-NIFK Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot:1.500-1.3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: NIFK (nucleolar protein interacting with the FHA domain of MKI67)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR