Anti-CABLES1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA315984EA 552 EUR
ORIGTA315984
Anti-CABLES1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CABLES1 (Cdk5 and Abl enzyme substrate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR