Anti-MOB3C Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316158EA 552 EUR
ORIGTA316158
Anti-MOB3C Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: MOB3C (MOB kinase activator 3C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR