Anti-OR56B4 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316210EA 532 EUR
ORIGTA316210
Anti-OR56B4 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: OR56B4 (olfactory receptor, family 56, subfamily B, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR