Anti-OR6Q1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316240EA 532 EUR
ORIGTA316240
Anti-OR6Q1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: OR6Q1 (olfactory receptor, family 6, subfamily Q, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR