Anti-PTRF Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316311EA 552 EUR
ORIGTA316311
Anti-PTRF Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:10000

Type: Primary
Antigen: PTRF (polymerase I and transcript release factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR