Anti-HKR1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316317EA 532 EUR
ORIGTA316317
Anti-HKR1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: HKR1 (HKR1, GLI-Kruppel zinc finger family member)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR