Anti-SERINC2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316384EA 552 EUR
ORIGTA316384
Anti-SERINC2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: SERINC2 (serine incorporator 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR