Anti-TBX10 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316385EA 552 EUR
ORIGTA316385
Anti-TBX10 Rabbit Polyclonal Antibody
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: TBX10 (T-box 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR