Anti-OR56A3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316415EA 552 EUR
ORIGTA316415
Anti-OR56A3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: OR56A3 (olfactory receptor, family 56, subfamily A, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR