Anti-ANK3 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316531EA 666 EUR
ORIGTA316531
Anti-ANK3 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 3.75 µg/ml

Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR