Anti-BEST4 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA316567EA 666 EUR
ORIGTA316567
Anti-BEST4 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: BEST4 (bestrophin 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR