Anti-MFGE8 Mouse Monoclonal Antibody [clone: B656M]

Toimittaja: OriGene
ORIGTA317012EA 666 EUR
ORIGTA317012
Anti-MFGE8 Mouse Monoclonal Antibody [clone: B656M]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: MFGE8 (milk fat globule-EGF factor 8 protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR