Anti-XCR1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA317533EA 666 EUR
ORIGTA317533
Anti-XCR1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: XCR1 (chemokine (C motif) receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR