Anti-SPP1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA318038EA 666 EUR
ORIGTA318038
Anti-SPP1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Immunochemistry: 5 µg/ml; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR