Anti-FGFR1 Mouse Monoclonal Antibody [clone: M 17A3]

Toimittaja: OriGene
ORIGTA318605EA 771 EUR
ORIGTA318605
Anti-FGFR1 Mouse Monoclonal Antibody [clone: M 17A3]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 10 µg/ml; Immunoprecipitation; Western Blot: 1 µg/ml

Type: Primary
Antigen: FGFR1 (fibroblast growth factor receptor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR