Anti-MDH2 Mouse Monoclonal Antibody [clone: 1G12]

Toimittaja: OriGene
ORIGTA318626EA 867 EUR
ORIGTA318626
Anti-MDH2 Mouse Monoclonal Antibody [clone: 1G12]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 10 µg/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: MDH2 (malate dehydrogenase 2, NAD (mitochondrial))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR