Anti-TLR5 Rabbit Monoclonal Antibody

Toimittaja: OriGene
ORIGTA318659EA 666 EUR
ORIGTA318659
Anti-TLR5 Rabbit Monoclonal Antibody
Vasta-aineet
Recommended Dilutions: Flo; Immunochemistry: 10 µg/ml; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: TLR5 (toll-like receptor 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR