Anti-ENTPD2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA318850EA 666 EUR
ORIGTA318850
Anti-ENTPD2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 15-20 µg/ml

Type: Primary
Antigen: ENTPD2 (ectonucleoside triphosphate diphosphohydrolase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR