Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI4G3]

Toimittaja: OriGene
ORIGTA500042EA 608 EUR
ORIGTA500042
Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI4G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2500-5000; Immunochemistry: 1:50

Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR