Anti-TUBB3 Mouse Monoclonal Antibody [clone: OTI5H2]

Toimittaja: OriGene
ORIGTA500047EA 621 EUR
ORIGTA500047
Anti-TUBB3 Mouse Monoclonal Antibody [clone: OTI5H2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR