Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI2A3]

Toimittaja: OriGene
ORIGTA500053EA 581 EUR
ORIGTA500053
Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI2A3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR