Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI2H9]

Toimittaja: OriGene
ORIGTA500056EA 608 EUR
ORIGTA500056
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI2H9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR