Anti-PROM2 Mouse Monoclonal Antibody [clone: OTI13A9]

Toimittaja: OriGene
ORIGTA500350EA 581 EUR
ORIGTA500350
Anti-PROM2 Mouse Monoclonal Antibody [clone: OTI13A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PROM2 (prominin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR