Anti-CDK5 Mouse Monoclonal Antibody [clone: OTI2H6]

Toimittaja: OriGene
ORIGTA500397EA 608 EUR
ORIGTA500397
Anti-CDK5 Mouse Monoclonal Antibody [clone: OTI2H6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: CDK5 (cyclin-dependent kinase 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR