Anti-MAP2K4 Mouse Monoclonal Antibody [clone: OTI4B4]

Toimittaja: OriGene
ORIGTA500411EA 603 EUR
ORIGTA500411
Anti-MAP2K4 Mouse Monoclonal Antibody [clone: OTI4B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR