Anti-PARP16 Mouse Monoclonal Antibody [clone: OTI2C8]

Toimittaja: OriGene
ORIGTA500452EA 608 EUR
ORIGTA500452
Anti-PARP16 Mouse Monoclonal Antibody [clone: OTI2C8]
Vasta-aineet
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: PARP16 (poly (ADP-ribose) polymerase family, member 16)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR