Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI9E9]

Toimittaja: OriGene
ORIGTA500467EA 603 EUR
ORIGTA500467
Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI9E9]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunoprecipitation: 2μg/500μl

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR