Anti-MRI1 Mouse Monoclonal Antibody [clone: OTI9H7]

Toimittaja: OriGene
ORIGTA500491EA 603 EUR
ORIGTA500491
Anti-MRI1 Mouse Monoclonal Antibody [clone: OTI9H7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MRI1 (methylthioribose-1-phosphate isomerase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR