Anti-FERMT2 Mouse Monoclonal Antibody [clone: OTI14A11]

Toimittaja: OriGene
ORIGTA500505EA 603 EUR
ORIGTA500505
Anti-FERMT2 Mouse Monoclonal Antibody [clone: OTI14A11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FERMT2 (fermitin family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR