Anti-CTAG1B Mouse Monoclonal Antibody [clone: OTI3C9]

Toimittaja: OriGene
ORIGTA500519EA 603 EUR
ORIGTA500519
Anti-CTAG1B Mouse Monoclonal Antibody [clone: OTI3C9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: CTAG1B (cancer/testis antigen 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR